4-Benzyloxy-2-nitrotoluene

Leverandør: Apollo Scientific

OR470680-5G
APOSOR470680-5GEA 4900 NOK
APOSOR470680-5G
4-Benzyloxy-2-nitrotoluene
4-Benzyloxy-2-nitrotoluene
Formel: C₁₄H₁₃NO₃
MW: 243,258 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00100651
CAS-nummer: 24239-67-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR