5-Bromo-N,N-diethyl-1-pentanamine hydrobromide

Leverandør: Apollo Scientific

OR470746-1G
APOSOR470746-1GEA 13775 NOK
APOSOR470746-1G
5-Bromo-N,N-diethyl-1-pentanamine hydrobromide
5-Bromo-N,N-diethyl-1-pentanamine hydrobromide
Formel: C₉H₂₀BrN·BrH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD27500643
CAS-nummer: 2006277-83-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR