4-Bromo-N,N-diethyl-1-butanamine hydrobromide

Leverandør: Apollo Scientific

OR470748-1G
APOSOR470748-1GEA 9900 NOK
APOSOR470748-1G
4-Bromo-N,N-diethyl-1-butanamine hydrobromide
4-Bromo-N,N-diethyl-1-butanamine hydrobromide
Formel: C₈H₁₈BrN·BrH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21602539
CAS-nummer: 856988-73-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR