Benzyl N-Boc-azetidine-3-carboxylate

Leverandør: Apollo Scientific

OR470841-5G
APOSOR470841-5GEA 7275 NOK
APOSOR470841-5G
Benzyl N-Boc-azetidine-3-carboxylate
Benzyl N-Boc-azetidine-3-carboxylate
Formel: C₁₆H₂₁NO₄
MW: 291.342 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD26936044
CAS-nummer: 1803599-76-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR