4-Aza-9-fluorenone

Leverandør: Apollo Scientific

OR470948-1G
APOSOR470948-1GEA 5900 NOK
APOSOR470948-1G
4-Aza-9-fluorenone
4-Aza-9-fluorenone
Formel: C₁₂H₇NO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00022194
CAS-nummer: 3882-46-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR