3-Amino-3-methyl-1-butanol hydrochloride

Leverandør: Apollo Scientific

OR471003-5G
APOSOR471003-5GEA 9900 NOK
APOSOR471003-5G
3-Amino-3-methyl-1-butanol hydrochloride
3-Amino-3-methyl-1-butanol hydrochloride
Formel: C₅H₁₃NO·ClH
MW: 139,624 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD26407724
CAS-nummer: 357185-97-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR