4-(Chloromethyl)piperidine hydrochloride

Leverandør: Apollo Scientific

OR471034-5G
APOSOR471034-5GEA 13775 NOK
APOSOR471034-5G
4-(Chloromethyl)piperidine hydrochloride
4-(Chloromethyl)piperidine hydrochloride
Formel: C₆H₁₂ClN·ClH
MW: 170.08 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11110948
CAS-nummer: 1822-61-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR