Methyl 3-(1-Hydroxyethyl)benzoate

Leverandør: Apollo Scientific

OR471065-1G
APOSOR471065-1GEA 8775 NOK
APOSOR471065-1G
Methyl 3-(1-Hydroxyethyl)benzoate
Methyl 3-(1-Hydroxyethyl)benzoate
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28122816
CAS-nummer: 87808-11-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR