1-Acetyl-4-(4-bromophenyl)piperazine

Leverandør: Apollo Scientific

OR471089-5G
APOSOR471089-5GEA 15150 NOK
APOSOR471089-5G
1-Acetyl-4-(4-bromophenyl)piperazine
1-Acetyl-4-(4-bromophenyl)piperazine
Formel: C₁₂H₁₅BrN₂O
MW: 283,164 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD17170409
CAS-nummer: 678996-43-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR