N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide

Leverandør: Apollo Scientific

OR471134-5G
APOSOR471134-5GEA 7400 NOK
APOSOR471134-5G
N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide
N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide
Formel: C₁₁H₉N₃O₃S
MW: 263.272 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01023820
CAS-nummer: 53173-92-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR