2-Hydroxy-3-methylbenzamide

Leverandør: Apollo Scientific

OR471175-1G
APOSOR471175-1GEA 7025 NOK
APOSOR471175-1G
2-Hydroxy-3-methylbenzamide
2-Hydroxy-3-methylbenzamide
Formel: C₈H₉NO₂
MW: 151,163 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00242811
CAS-nummer: 14008-60-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR