Quinoline-4-carbohydrazide Hydrate

Leverandør: Apollo Scientific

OR471337-1G OR471337-250MG
APOSOR471337-1GEA 12400 NOK
APOSOR471337-1G APOSOR471337-250MG
Quinoline-4-carbohydrazide Hydrate
Quinoline-4-carbohydrazide Hydrate
Formel: C₁₀H₉N₃O·H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28968740
CAS-nummer: 2006277-54-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR