Ethyl 3-Ethylisoxazole-4-carboxylate

Leverandør: Apollo Scientific

OR471415-5G
APOSOR471415-5GEA 9900 NOK
APOSOR471415-5G
Ethyl 3-Ethylisoxazole-4-carboxylate
Ethyl 3-Ethylisoxazole-4-carboxylate
Formel: C₈H₁₁NO₃
MW: 169,178 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18833193
CAS-nummer: 639523-11-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR