2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Leverandør: Apollo Scientific

Synonymer: 4-Hydroxyphenylboronic acid MIDA ester

OR471520-5G
APOSOR471520-5GEA 14850 NOK
APOSOR471520-5G
2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Formel: C₁₁H₁₂BNO₅
MW: 249,028 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD25541468
CAS-nummer: 1613586-40-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR