3-Amino-3-(2-naphthyl)propionic acid 95%

Leverandør: Apollo Scientific

Synonymer: DL-3-Amino-3-(2-naphthyl)propionic acid

OR471651-25G
APOSOR471651-25GEA 2875 NOK
APOSOR471651-25G
3-Amino-3-(2-naphthyl)propionic acid 95%
3-Amino-3-(2-naphthyl)propionic acid
Formel: C₁₃H₁₃NO₂
MW: 215.248 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01871322
CAS-nummer: 129042-57-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR