4-Bromopiperidine hydrobromide 98%

Leverandør: Apollo Scientific

OR480694-25G
APOSOR480694-25GEA 1125 NOK
APOSOR480694-25G
4-Bromopiperidine hydrobromide 98%
4-Bromopiperidine hydrobromide
Formel: C₅H₁₁Br₂N
MW: 244.9600 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00191858
CAS-nummer: 54288-70-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR