3-Chloro-4-cyanobenzenesulphonyl chloride 95%

Leverandør: Apollo Scientific

Synonymer: 3-Chloro-4-cyanobenzene-1-sulphonyl chloride

OR480708-5G
APOSOR480708-5GEA 12350 NOK
APOSOR480708-5G
3-Chloro-4-cyanobenzenesulphonyl chloride 95%
3-Chloro-4-cyanobenzenesulphonyl chloride
Formel: C₇H₃Cl₂NO₂S
MW: 236,075 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09812743
CAS-nummer: 213130-43-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR