5-Bromo-1H-indole-3-carboxylic acid 94%

Leverandør: Apollo Scientific

Synonymer: 5-Bromoindole-3-carboxylic acid

OR480712-25G
APOSOR480712-25GEA 2475 NOK
APOSOR480712-25G
5-Bromo-1H-indole-3-carboxylic acid 94%
5-Bromo-1H-indole-3-carboxylic acid
Formel: C₉H₆BrNO₂
MW: 240.06 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05664007
CAS-nummer: 10406-06-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR