2-(4-Hydroxybenzoyl)benzoic acid

Leverandør: Apollo Scientific
Warning

OR4859-5G OR4859-25G OR4859-100G
APOSOR4859-5GEA 270 NOK
APOSOR4859-5G APOSOR4859-25G APOSOR4859-100G
2-(4-Hydroxybenzoyl)benzoic acid
2-(4-Hydroxybenzoyl)benzoic acid
Formel: C₁₄H₁₀O₄
MW: 242.227 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016507
CAS-nummer: 85-57-4
EINECS: 201-616-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR