4,4'-Methylenedi-o-toluidine

Leverandør: Apollo Scientific

OR49083-25G
APOSOR49083-25GEA 735 NOK
APOSOR49083-25G
4,4'-Methylenedi-o-toluidine
4,4'-Methylenedi-o-toluidine
Formel: C₁₅H₁₈N₂
Smeltepunkt: 156 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00126963
CAS-nummer: 838-88-0
FN: 3077
ADR: 9+EHS,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR