5,8-Dichloroisoquinoline ≥97%

Leverandør: Apollo Scientific

OR510025-250MG
APOSOR510025-250MGEA 915 NOK
APOSOR510025-250MG
5,8-Dichloroisoquinoline ≥97%
5,8-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
MW: 198,049 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD07644632
CAS-nummer: 73075-59-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR