5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine ≥97%

Leverandør: Apollo Scientific

Synonymer: 5,8-Dibromo[1,2,4]triazolo[1,5-a]pyrazine

OR510080-250MG
APOSOR510080-250MGEA 2300 NOK
APOSOR510080-250MG
5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine ≥97%
5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine
Formel: C₅H₂Br₂N₄
MW: 277,907 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11040378
CAS-nummer: 959755-46-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR