6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid ≥97%

Leverandør: Apollo Scientific

OR510105-250MG
APOSOR510105-250MGEA 1775 NOK
APOSOR510105-250MG
6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid ≥97%
6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid
Formel: C₇H₄BrN₃O₂
MW: 242,03 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 907945-69-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR