3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine ≥98%

Leverandør: Apollo Scientific

OR510175-100MG
APOSOR510175-100MGEA 5225 NOK
APOSOR510175-100MG
3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine ≥98%
3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine
Formel: C₅H₂BrClN₄
MW: 233.453 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 1936225-21-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR