2-Bromo-5,6-dichloro-1H-benzo[d]imidazole ≥98%

Leverandør: Apollo Scientific

OR510208-1G
APOSOR510208-1GEA 2000 NOK
APOSOR510208-1G
2-Bromo-5,6-dichloro-1H-benzo[d]imidazole ≥98%
2-Bromo-5,6-dichloro-1H-benzo[d]imidazole
Formel: C₇H₃BrCl₂N₂
MW: 265,922 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 142356-40-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR