2,2':5',2''-Terthiophene 98%

Leverandør: Apollo Scientific
Warning

OR51731-25G
APOSOR51731-25GEA 1200 NOK
APOSOR51731-25G
2,2':5',2''-Terthiophene 98%
2,2':5',2''-Terthiophene
Formel: C₁₂H₈S₃
MW: 248.39 g/mol
Kokepunkt: 160 °C/0.1mmHg
Smeltepunkt: 93…96 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012167
CAS-nummer: 1081-34-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR