Gallussyre, vannfri 99%

Leverandør: Apollo Scientific
Warning

OR51754-1KG OR51754-250G OR51754-5KG
APOSOR51754-1KGEA 1175 NOK
APOSOR51754-1KG APOSOR51754-250G APOSOR51754-5KG
Gallussyre, vannfri 99%
Gallussyre
Formel: C₇H₆O₅
Smeltepunkt: 251 °C (dec.)
Tetthet: 1,694 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002510
CAS-nummer: 149-91-7
EINECS: 205-749-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR