6-Nitro-m-toluidine

Leverandør: Apollo Scientific
Danger

OR51790-25G OR51790-5G
APOSOR51790-25GEA 1025 NOK
APOSOR51790-25G APOSOR51790-5G
6-Nitro-m-toluidine
6-Nitro-m-toluidine
Formel: C₇H₈N₂O₂
Smeltepunkt: 110…111 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007814
CAS-nummer: 578-46-1
EINECS: 209-423-7
FN: 2660
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR