p-Phenylenediamine dihydrochloride 99%

Leverandør: Apollo Scientific
Danger

OR51807-500G
APOSOR51807-500GEA 3800 NOK
APOSOR51807-500G
p-Phenylenediamine dihydrochloride 99%
p-Phenylenediamine dihydrochloride
Formel: C₆H₁₀Cl₂N₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013002
CAS-nummer: 624-18-0
FN: 1673
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR