(S)-(-)-Malein syre 99%

Leverandør: Apollo Scientific
Warning

OR51833-2.5KG OR51833-10KG
APOSOR51833-2.5KGEA 3250 NOK
APOSOR51833-2.5KG APOSOR51833-10KG
(S)-(-)-Malein syre 99%
(S)-(-)-Malein syre
Formel: C₄H₆O₅
Smeltepunkt: 101…103 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064213
CAS-nummer: 97-67-6
EINECS: 202-601-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR