(-)-3-[(3S,4S)-3,4-Dimethylpiperidin-4-yl]phenol 95%

Leverandør: Apollo Scientific
Warning

Synonymer: (3S,4S)-(-)-3,4-Dimethyl-4-(3-hydroxyphenyl)piperidine

OR51856-250MG
APOSOR51856-250MGEA 5025 NOK
APOSOR51856-250MG
(-)-3-[(3S,4S)-3,4-Dimethylpiperidin-4-yl]phenol 95%
(-)-3-[(3S,4S)-3,4-Dimethylpiperidin-4-yl]phenol
Formel: C₁₃H₁₉NO
MW: 205,296 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23134657
CAS-nummer: 145678-87-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR