m-Toluidin 99%

Leverandør: Apollo Scientific
Danger

Synonymer: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline

OR5362-250G OR5362-1KG
APOSOR5362-250GEA 1075 NOK
APOSOR5362-250G APOSOR5362-1KG
m-Toluidin 99%
m-Toluidin
Formel: C₇H₉N
Kokepunkt: 203…204 °C
Smeltepunkt: –30 °C
Tetthet: 0,999 g/cm³ (25 °C)
Flash punkt: 85 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007808
CAS-nummer: 108-44-1
EINECS: 203-583-1
FN: 1708
ADR: 6.1,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR