4-Methyl-2-nitrophenyl isocyanate 97%

Leverandør: Apollo Scientific
Danger

OR54499-1G OR54499-5G
APOSOR54499-1GEA 4300 NOK
APOSOR54499-1G APOSOR54499-5G
4-Methyl-2-nitrophenyl isocyanate 97%
4-Methyl-2-nitrophenyl isocyanate

When melted, may contain small amount of sediment.

Formel: C₈H₆N₂O₃
MW: 178.15 g/mol
Smeltepunkt: 56…58 °C
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014705
CAS-nummer: 57910-98-2
FN: 3335
ADR: 9 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR