4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde

Leverandør: Apollo Scientific
Warning

OR54639-1G
APOSOR54639-1GEA 1750 NOK
APOSOR54639-1G
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
Formel: C₁₀H₈N₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07772818
CAS-nummer: 852180-60-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR