(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95+%

Leverandør: Apollo Scientific
Warning

Synonymer: (S)-2,2'-((5-Amino-1-carboxypentyl)azanediyl)diacetic acid, Nα,Nα-Bis(carboxymethyl)-L-lysine

OR55124-5G
APOSOR55124-5GEA 1200 NOK
APOSOR55124-5G
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95+%
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid
Formel: C₁₀H₁₈N₂O₆
MW: 262.26 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00237437
CAS-nummer: 113231-05-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR