Quinoline-N-oxide hydrate

Leverandør: Apollo Scientific
Warning

Synonymer: Quinoline 1-oxide hydrate

OR55323-25G OR55323-5G
APOSOR55323-25GEA 3600 NOK
APOSOR55323-25G APOSOR55323-5G
Quinoline-N-oxide hydrate
Quinoline-N-oxide hydrate
Formel: C₉H₇NO·xH₂O
Smeltepunkt: 52…55 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149472
CAS-nummer: 64201-64-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR