1,3-Dihydroksynaftalen 99%

Leverandør: Apollo Scientific
Warning

OR55332-1G OR55332-25G OR55332-5G
APOSOR55332-1GEA 450 NOK
APOSOR55332-1G APOSOR55332-25G APOSOR55332-5G
1,3-Dihydroksynaftalen 99%
1,3-Dihydroksynaftalen
Formel: C₁₀H₈O₂
Smeltepunkt: 123…126 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003965
CAS-nummer: 132-86-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR