1-Boc-4-chlorosulphonylpiperidine 98%

Leverandør: Apollo Scientific
Danger

Synonymer: 1-Boc-4-Chlorosulphonylpiperidine, 1-BOC-4-PIPERIDINEsulphONYL CHLORIDE, tert-Butyl 4-(chlorosulphonyl)piperidine-1-carboxylate, 4-CHLOROsulphONYL-PIPERIDINE-1-CARBOXYLIC acid TERT-BUTYL ESTER

OR55455-5G
APOSOR55455-5GEA 2150 NOK
APOSOR55455-5G
1-Boc-4-chlorosulphonylpiperidine 98%
1-Boc-4-chlorosulphonylpiperidine
Formel: C₁₀H₁₈ClNO₄S
MW: 283.767 g/mol
Smeltepunkt: 82…85 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01861750
CAS-nummer: 782501-25-1
FN: 3261
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR