4,5-Dibromo-1H-pyrrole-2-carboxaldehyde 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 2,3-Dibromo-5-formyl-1H-pyrrole

OR5763-1G
APOSOR5763-1GEA 3800 NOK
APOSOR5763-1G
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde 98%
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde
Formel: C₅H₃Br₂NO
MW: 252,891 g/mol
Smeltepunkt: 154…158 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD07363813
CAS-nummer: 932-82-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR