2,5-Dimethoxyphenylboronic acid

Leverandør: Apollo Scientific
Warning

Synonymer: (2,5-Dimethoxyphenyl)metaboric acid, 2,5-Dimethoxybenzeneboronic acid, (2,5-Dimethoxyphenyl)boronic acid, Dihydroxy(2,5-dimethoxyphenyl)borane

OR5823-100G
APOSOR5823-100GEA 2425 NOK
APOSOR5823-100G
2,5-Dimethoxyphenylboronic acid
2,5-Dimethoxyphenylboronic acid
Formel: C₈H₁₁BO₄
MW: 181.98 g/mol
Smeltepunkt: 95…98 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01318181
CAS-nummer: 107099-99-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR