N,N-Diethyl-p-toluidine

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Diethylaminotoluene, N,N-Diethyl-4-methylaniline

OR5841-25G OR5841-5G
APOSOR5841-25GEA 1175 NOK
APOSOR5841-25G APOSOR5841-5G
N,N-Diethyl-p-toluidine
N,N-Diethyl-p-toluidine
Formel: C₁₁H₁₇N
Kokepunkt: 230 °C
Tetthet: 0,92 g/cm³ (25 °C)
Flash punkt: 102 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015113
CAS-nummer: 613-48-9
EINECS: 210-345-0
FN: 2810
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR