4'-Chloro-3'-hydroxyacetanilide

Leverandør: Apollo Scientific

Synonymer: N-(4-Chloro-3-hydroxyphenyl)acetamide

OR59372-1G
APOSOR59372-1GEA 2175 NOK
APOSOR59372-1G
4'-Chloro-3'-hydroxyacetanilide
4'-Chloro-3'-hydroxyacetanilide
Formel: C₈H₈ClNO₂
MW: 185,6100 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09878312
CAS-nummer: 28443-52-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR