(3,4-Dimethoxyphenyl)acetic acid 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Homoveratric acid, 3,4-Dimethoxyphenylacetic acid

OR5989-500G
APOSOR5989-500GEA 1300 NOK
APOSOR5989-500G
(3,4-Dimethoxyphenyl)acetic acid 98%
(3,4-Dimethoxyphenyl)acetic acid
Formel: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Kokepunkt: 327
Smeltepunkt: 97…99 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004335
CAS-nummer: 93-40-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR