Ethyl glyoxylate 50% in toluene, Technical Grade

Leverandør: Apollo Scientific
Danger

OR59968-100G OR59968-500G OR59968-25G
APOSOR59968-100GEA 715 NOK
APOSOR59968-100G APOSOR59968-500G APOSOR59968-25G
Ethyl glyoxylate 50% in toluene, Technical Grade
Ethyl glyoxylate
Formel: C₄H₆O₃
Kokepunkt: 110 °C
Tetthet: 1,003 g/cm³ (25 °C)
Flash punkt: 7 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044009
CAS-nummer: 924-44-7
FN: 1294
ADR: 3,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR