Glyoxal dimethyl acetal 60% in aqueous solution

Leverandør: Apollo Scientific
Warning

Synonymer: Dimethoxyacetaldehyde, 2,2-Dimethoxyacetaldehyde, Dimethoxyethanal

OR59985-100G OR59985-500G OR59985-5G
APOSOR59985-100GEA 985 NOK
APOSOR59985-100G APOSOR59985-500G APOSOR59985-5G
Glyoxal dimethyl acetal 60% in aqueous solution
Glyoxal dimethyl acetal
Formel: OHCCH(OCH₃)₂
Kokepunkt: 100…102 °C/759mmHg
Tetthet: 1,15 g/cm³ (25 °C)
Flash punkt: 77 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134410
CAS-nummer: 51673-84-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR