5-Isocyanato-1,3-benzodioxole 95%

Leverandør: Apollo Scientific
Warning

Synonymer: 1,3-Benzodioxol-5-yl isocyanate, 5-Isocyanatobenzo[d][1,3]dioxole

OR61306-1G OR61306-5G
APOSOR61306-1GEA 680 NOK
APOSOR61306-1G APOSOR61306-5G
5-Isocyanato-1,3-benzodioxole 95%
5-Isocyanato-1,3-benzodioxole

When melted, may contain small amount of sediment.

Formel: C₈H₅NO₃
MW: 163.13 g/mol
Smeltepunkt: 48…51 °C
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01863692
CAS-nummer: 69922-28-7
FN: 3335
ADR: 9 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR