6-Bromo-m-toluidine 98%

Leverandør: Apollo Scientific
Warning

OR61400-100G OR61400-25G OR61400-500G
APOSOR61400-100GEA 825 NOK
APOSOR61400-100G APOSOR61400-25G APOSOR61400-500G
6-Bromo-m-toluidine 98%
6-Bromo-m-toluidine
Formel: C₇H₈BrN
Smeltepunkt: 32…34 °C
Tetthet: 1,486 g/cm³ (25 °C)
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01806278
CAS-nummer: 53078-85-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR