4,6-Dichloro-o-toluidine 97%

Leverandør: Apollo Scientific
Warning

OR6295-1G OR6295-25G OR6295-5G
APOSOR6295-1GEA 270 NOK
APOSOR6295-1G APOSOR6295-25G APOSOR6295-5G
4,6-Dichloro-o-toluidine 97%
4,6-Dichloro-o-toluidine
Formel: C₇H₇Cl₂N
Kokepunkt: 82…84 °C/0,1mmHg
Smeltepunkt: 42…46 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044074
CAS-nummer: 30273-00-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR