Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%

Leverandør: Apollo Scientific

Synonymer: Methyl 3-bromo-2-oxo-2H-pyran-5-carboxylate

OR6402-5G
APOSOR6402-5GEA 1125 NOK
APOSOR6402-5G
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate
Formel: C₇H₅BrO₄
MW: 233,02 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00463742
CAS-nummer: 42933-07-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR