4-Methyl-3-nitrophenyl isocyanate 97%

Leverandør: Apollo Scientific
Danger

OR6434-10G OR6434-1G OR6434-5G
APOSOR6434-10GEA 2375 NOK
APOSOR6434-10G APOSOR6434-1G APOSOR6434-5G
4-Methyl-3-nitrophenyl isocyanate 97%
4-Methyl-3-nitrophenyl isocyanate

When melted, may contain small amount of sediment.

Formel: C₈H₆N₂O₃
MW: 178.15 g/mol
Kokepunkt: 113…118 °C/3mmHg
Smeltepunkt: 48…54 °C
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014698
CAS-nummer: 13471-69-7
FN: 2811
ADR: 6.1 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR